ENA394766885 Enamine

3-(4-chloro-3-fluorobenzoyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD24842800
  • Cas Number : 1443981-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127882
Availability details for
Quantity
  • Availability
  • Add to Cart