ENA394766887 Enamine

3-[4-(trifluoromethyl)benzoyl]-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD24842803
  • Cas Number : 1256257-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127887
Availability details for
Quantity
  • Availability
  • Add to Cart