ENA394766888 Enamine

1-(cyclopropylmethoxy)-4-fluoro-2-nitrobenzene

  • MDL number MFCD24842804
  • Cas Number : 1092496-28-0
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127908
Availability details for
Quantity
  • Availability
  • Add to Cart