ENA394766889 Enamine

methyl 6-[(cyclopropylformamido)methyl]-2-methylpyridine-3-carboxylate

  • MDL number MFCD24842806
  • Cas Number : 1443980-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127911
Availability details for
Quantity
  • Availability
  • Add to Cart