ENA394766893 Enamine

octahydropiperazino[2,1-c]morpholine-6,9-dione

  • MDL number MFCD24842810
  • Cas Number : 1443979-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127998
Availability details for
Quantity
  • Availability
  • Add to Cart