ENA394766894 Enamine

(7-chloroquinolin-8-yl)methanol

  • MDL number MFCD24842811
  • Cas Number : 84163-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128014

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128014
Availability details for
Quantity
  • Availability
  • Add to Cart