ENA394766895 Enamine

1,3-diethyl 4-oxopiperidine-1,3-dicarboxylate

  • MDL number MFCD24842812
  • Cas Number : 53601-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128016
Availability details for
Quantity
  • Availability
  • Add to Cart