ENA394766896 Enamine

5-cyano-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD24842813
  • Cas Number : 1443980-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128019
Availability details for
Quantity
  • Availability
  • Add to Cart