ENA394766901 Enamine

2-(adamantan-1-yl)-2-(morpholin-4-yl)acetic acid

  • Cas Number : 1443979-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128095
Availability details for
Quantity
  • Availability
  • Add to Cart