ENA394766903 Enamine

N'-(2-chloroacetyl)-2-methoxyacetohydrazide

  • MDL number MFCD24842820
  • Cas Number : 167155-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128099
Availability details for
Quantity
  • Availability
  • Add to Cart