ENA394766904 Enamine

3-(benzyloxy)-2-(tert-butoxy)pyridine

  • MDL number MFCD24842821
  • Cas Number : 1443980-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128101
Availability details for
Quantity
  • Availability
  • Add to Cart