ENA394766905 Enamine

4-(benzenesulfonyl)-1H-imidazole

  • MDL number MFCD24842822
  • Cas Number : 1443980-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128103
Availability details for
Quantity
  • Availability
  • Add to Cart