ENA394766909 Enamine

methyl 2-(2-bromo-2-methylpropanamido)-2-methylpropanoate

  • MDL number MFCD24842826
  • Cas Number : 151142-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128114
Availability details for
Quantity
  • Availability
  • Add to Cart