ENA394766910 Enamine

5-(chloromethyl)-1-(2-methylpropyl)-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD24842827
  • Cas Number : 1443980-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128125

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128125
Availability details for
Quantity
  • Availability
  • Add to Cart