ENA394766912 Enamine

methyl 4-(chlorosulfonyl)-5-ethylfuran-2-carboxylate

  • MDL number MFCD24842829
  • Cas Number : 1443981-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128184

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128184
Availability details for
Quantity
  • Availability
  • Add to Cart