ENA394766914 Enamine

tert-butyl 4-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)piperidine-1-carboxylate

  • MDL number MFCD24842832
  • Cas Number : 864293-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128191
Availability details for
Quantity
  • Availability
  • Add to Cart