ENA394766923 Enamine

1,3-dimethyl 2-(furan-2-ylmethyl)propanedioate

  • MDL number MFCD24842845
  • Cas Number : 99187-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128240

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128240
Availability details for
Quantity
  • Availability
  • Add to Cart