ENA394766927 Enamine

3-methyl-2-(2-methylpiperidin-1-yl)-5-nitropyridine

  • MDL number MFCD24842854
  • Cas Number : 1443982-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48...50
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128311

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128311
Availability details for
Quantity
  • Availability
  • Add to Cart