ENA394766931 Enamine

[1-(4-benzylmorpholin-2-yl)ethyl](methyl)amine

  • MDL number MFCD24842858
  • Cas Number : 1443981-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128372
Availability details for
Quantity
  • Availability
  • Add to Cart