ENA394766936 Enamine

benzyl[1-(2,6-difluorophenyl)-2,2,2-trifluoroethyl]amine

  • MDL number MFCD24842863
  • Cas Number : 1443981-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128397
Availability details for
Quantity
  • Availability
  • Add to Cart