ENA394766941 Enamine

1-[4-benzyl-3-(propan-2-yl)piperazin-1-yl]-2,2,2-trifluoroethan-1-one

  • MDL number MFCD24842870
  • Cas Number : 1443980-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128434
Availability details for
Quantity
  • Availability
  • Add to Cart