ENA394766942 Enamine

3-[methyl(pyridin-4-yl)amino]propanoic acid hydrochloride

  • MDL number MFCD24842871
  • Cas Number : 193818-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128443

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-128443
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart