ENA394766947 Enamine

phenyl N-(5-methyl-1,2,4-oxadiazol-3-yl)carbamate

  • MDL number MFCD24842876
  • Cas Number : 1182358-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128577
Availability details for
Quantity
  • Availability
  • Add to Cart