ENA394766953 Enamine

2-amino-5,5,5-trifluoropentanoic acid hydrochloride

  • MDL number MFCD24842885
  • Cas Number : 1443979-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128644
Availability details for
Quantity
  • Availability
  • Add to Cart