ENA394766954 Enamine

1,1-dimethoxycyclobutane

  • MDL number MFCD24842886
  • Cas Number : 4415-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128645

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128645
Availability details for
Quantity
  • Availability
  • Add to Cart