ENA394766956 Enamine

3-propyl-1-[3-(trifluoromethyl)phenyl]piperazine hydrochloride

  • MDL number MFCD24842888
  • Cas Number : 1443980-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128653
Availability details for
Quantity
  • Availability
  • Add to Cart