ENA394766964 Enamine

pent-4-en-1-yl 2-{[(tert-butoxy)carbonyl]amino}acetate

  • MDL number MFCD24394354
  • Cas Number : 1356164-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128747
Availability details for
Quantity
  • Availability
  • Add to Cart