ENA394766967 Enamine

4-[4-(hydroxymethyl)-1-methyl-1H-pyrazol-3-yl]benzonitrile

  • MDL number MFCD24842903
  • Cas Number : 1443981-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128864
Availability details for
Quantity
  • Availability
  • Add to Cart