ENA394766969 Enamine

1-(oxan-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD24842905
  • Cas Number : 1443981-91-6
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tetrahydro-2H-pyran-3-yl)ethan-1-amine hydrochloride
Inchi Code 1S/C7H15NO.ClH/c1-6(8)7-3-2-4-9-5-7;/h6-7H,2-5,8H2,1H3;1H
Inchi Key CPHBDOYIYBNGNN-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128882
Availability details for
Quantity
  • Availability
  • Add to Cart