ENA394766971 Enamine

piperidin-4-yl methanesulfonate hydrochloride

  • MDL number MFCD24842907
  • Cas Number : 1443980-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222...224
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128939
Availability details for
Quantity
  • Availability
  • Add to Cart