ENA394766977 Enamine

1,3-diethyl 2-(2-chloro-4-nitrobenzoyl)propanedioate

  • MDL number MFCD24842920
  • Cas Number : 1443979-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129055
Availability details for
Quantity
  • Availability
  • Add to Cart