ENA394766981 Enamine

6-methoxy-1,3-dimethyl-3,4-dihydroisoquinoline

  • MDL number MFCD24842925
  • Cas Number : 1443980-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129079
Availability details for
Quantity
  • Availability
  • Add to Cart