ENA394766987 Enamine

(1S)-1-phenylpentan-1-amine hydrochloride

  • MDL number MFCD24842931
  • Cas Number : 911373-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129108
Availability details for
Quantity
  • Availability
  • Add to Cart