ENA394766998 Enamine

tert-butyl 2-cyano-2,2-dimethylacetate

  • MDL number MFCD24842949
  • Cas Number : 75235-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129289

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129289
Availability details for
Quantity
  • Availability
  • Add to Cart