ENA394767011 Enamine

methyl 2-(3-cyanophenyl)-1,3-oxazole-4-carboxylate

  • MDL number MFCD24842961
  • Cas Number : 1239589-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129321
Availability details for
Quantity
  • Availability
  • Add to Cart