ENA394767012 Enamine

6-hydroxy-5-sulfopyridine-3-carboxylic acid

  • MDL number MFCD24842962
  • Cas Number : 1443980-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129338
Availability details for
Quantity
  • Availability
  • Add to Cart