ENA394767013 Enamine

2-(2,4-difluorophenyl)ethanimidamide hydrochloride

  • MDL number MFCD24842963
  • Cas Number : 139161-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129343

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129343
Availability details for
Quantity
  • Availability
  • Add to Cart