ENA394767016 Enamine

tert-butyl N-(3,5-dimethylpyridin-2-yl)carbamate

  • MDL number MFCD24842966
  • Cas Number : 1237069-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129375
Availability details for
Quantity
  • Availability
  • Add to Cart