ENA394767018 Enamine

1-(2,6-difluorophenyl)-2,2,2-trifluoroethan-1-amine hydrochloride

  • MDL number MFCD24842970
  • Cas Number : 1443981-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129421

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129421
Availability details for
Quantity
  • Availability
  • Add to Cart