ENA394767023 Enamine

tert-butyl 4-methyl-4-(3-methyl-1,2,4-oxadiazol-5-yl)piperidine-1-carboxylate

  • MDL number MFCD24842980
  • Cas Number : 1443981-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129457
Availability details for
Quantity
  • Availability
  • Add to Cart