ENA394767041 Enamine

1-(2-methoxyethyl)-3-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD24842762
  • Cas Number : 1443981-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125808

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125808
Availability details for
Quantity
  • Availability
  • Add to Cart