ENA394767071 Enamine

4-(tert-butoxy)-2-cyano-2-methyl-4-oxobutanoic acid

  • MDL number MFCD24450179
  • Cas Number : 1443980-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127112

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127112
Availability details for
Quantity
  • Availability
  • Add to Cart