ENA394767073 Enamine

1-(1,3-thiazol-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16065431
  • Cas Number : 1247644-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127127

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-127127
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart