ENA394767082 Enamine

(1R)-1-(2-bromo-4-methoxyphenyl)ethan-1-ol

  • MDL number MFCD24302926
  • Cas Number : 1232403-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127317
Availability details for
Quantity
  • Availability
  • Add to Cart