ENA394767086 Enamine

ethyl [1,3]oxazolo[4,5-b]pyridine-2-carboxylate

  • MDL number MFCD13195427
  • Cas Number : 93129-56-7
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl [1,3]oxazolo[4,5-b]pyridine-2-carboxylate
Inchi Code 1S/C9H8N2O3/c1-2-13-9(12)8-11-7-6(14-8)4-3-5-10-7/h3-5H,2H2,1H3
Inchi Key KRPJHFZZVZJQES-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127422

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-127422
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart