ENA394767103 Enamine

4-bromo-1-(2-methoxyethoxy)-2-nitrobenzene

  • MDL number MFCD14652110
  • Cas Number : 1248106-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127681
Availability details for
Quantity
  • Availability
  • Add to Cart