ENA394767106 Enamine

2-(tert-butylsulfanyl)pyridine-4-carboxylic acid

  • MDL number MFCD11136038
  • Cas Number : 1019539-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127705
Availability details for
Quantity
  • Availability
  • Add to Cart