ENA394767107 Enamine

2-hydroxybutanamide

  • MDL number MFCD16694120
  • Cas Number : 1113-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127708
Availability details for
Quantity
  • Availability
  • Add to Cart