ENA394767111 Enamine

2-fluoro-3-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD11036242
  • Cas Number : 1092349-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127745

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127745
Availability details for
Quantity
  • Availability
  • Add to Cart