ENA394767121 Enamine

1-(5-fluoropyridin-2-yl)propan-1-one

  • MDL number MFCD22724373
  • Cas Number : 1443980-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127773
Availability details for
Quantity
  • Availability
  • Add to Cart